Learn More
Medchemexpress LLC Thalidomide-5-COOH | 1216805-11-6 | 99.9% | 302.24 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY139539500MG
Encompass_Preferred
Thalidomide-5-COOH is a Thalidomide-based cereblon ligand utilized for the recruitment of CRBN protein. It can be linked to a protein ligand via a linker to form PROTACs.
  • Utilized for the recruitment of CRBN protein
  • Can be linked to a protein ligand via a linker
  • Forms PROTACs
  • PROTACs use the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-50881