Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide 5-fluoride | 835616-61-0 | 100.0% | 276.22 | C13H9FN2O4 | 1 ML
SDP

Catalog No. 5000208409
Encompass_Preferred

Thalidomide 5-fluoride is a thalidomide-derived cereblon ligand used as a building block in targeted protein degradation (PROTAC) research and related chemical biology applications. It is supplied as both solid material and pre-formulated DMSO solutions to facilitate linker conjugation and assay use.

  • Thalidomide-based cereblon ligand for CRBN recruitment.
  • Cas number: 835616-61-0.
  • Molecular formula C13H9FN2O4; molecular weight 276.22 g/mol.
  • Purity reported at 99.96% (analytical).
  • Available as 10 mM solution in DMSO (1 mL) and as solid in multiple package sizes.
  • Storage: powder at -20°C or 4°C; in solvent at -80°C or -20°C.

Catalog No. 50-002-08409 Supplier Medchemexpress LLC Supplier No. HYW08738310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Thalidomide 5-fluoride is a thalidomide-derived cereblon ligand used as a building block in targeted protein degradation (PROTAC) research and related chemical biology applications. It is supplied as both solid material and pre-formulated DMSO solutions to facilitate linker conjugation and assay use.

  • Thalidomide-based cereblon ligand for CRBN recruitment.
  • Cas number: 835616-61-0.
  • Molecular formula C13H9FN2O4; molecular weight 276.22 g/mol.
  • Purity reported at 99.96% (analytical).
  • Available as 10 mM solution in DMSO (1 mL) and as solid in multiple package sizes.
  • Storage: powder at -20°C or 4°C; in solvent at -80°C or -20°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.