Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide 5-fluoride | 835616-61-0 | 100.0% | 276.22 g/mol | C13H9FN2O4 | 25 G
SDP

Catalog No. 5000209453
Encompass_Preferred

Thalidomide 5-fluoride is a thalidomide-derived cereblon ligand used in targeted protein degradation research and PROTAC synthesis. It is supplied as a high-purity solid and as a DMSO solution, with analytical data and safety documentation available.

  • Thalidomide-based cereblon ligand for PROTAC development and E3 ligase recruitment studies.
  • Molecular formula C13H9FN2O4; molecular weight 276.22 g/mol.
  • High purity (99.96%).
  • Available as solid in 1 g, 5 g, 10 g, 25 g quantities.
  • Solution option available as 10 mM in DMSO.
  • Supplied with supporting documentation including COA, SDS, HNMR, RP-HPLC, and MS.

Catalog No. 50-002-09453 Supplier Medchemexpress LLC Supplier No. 25GHYW087383
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide 5-fluoride is a thalidomide-derived cereblon ligand used in targeted protein degradation research and PROTAC synthesis. It is supplied as a high-purity solid and as a DMSO solution, with analytical data and safety documentation available.

  • Thalidomide-based cereblon ligand for PROTAC development and E3 ligase recruitment studies.
  • Molecular formula C13H9FN2O4; molecular weight 276.22 g/mol.
  • High purity (99.96%).
  • Available as solid in 1 g, 5 g, 10 g, 25 g quantities.
  • Solution option available as 10 mM in DMSO.
  • Supplied with supporting documentation including COA, SDS, HNMR, RP-HPLC, and MS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.