Learn More
Medchemexpress LLC Thalidomide 5-fluoride | 835616-61-0 | MFCD30188075 | 100.0% | 276.22 g/mol | C13H9FN2O4 | 1 G

Supplier: Medchemexpress LLC HYW0873831G
Thalidomide 5-fluoride is a thalidomide-derived cereblon ligand used as an E3 ligase recruiting moiety in chemical biology and PROTAC research. It is supplied as a powder and as a 10 mM DMSO solution for laboratory use.
- High purity: 99.96%
- Molecular weight: 276.22 g/mol
- Chemical formula: C13H9FN2O4
- Available sizes: 1 mL (10 mM in DMSO), 1 G, 5 G, 10 G, 25 G, 50 G
- Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 6 months) or -20°C (up to 1 month)
- Safety and quality documents: SDS and COA provided by manufacturer
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.