Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide 5-fluoride | 835616-61-0 | MFCD30188075 | 100.0% | 276.22 g/mol | C13H9FN2O4 | 10 G
SDP

Catalog No. 5000208364
Encompass_Preferred

Thalidomide 5-fluoride is a thalidomide-derived cereblon ligand used as a building block in PROTAC degraders and other bifunctional molecules. It is supplied as a high-purity solid reagent intended for research and chemical biology applications.

  • High purity solid (99.96%).
  • Molecular weight 276.22 g/mol.
  • Molecular formula C13H9FN2O4.
  • Available in laboratory pack sizes, including 10 G.
  • Storage: powder at -20°C for long-term, 4°C for shorter term; in solvent store at -80°C.

Catalog No. 50-002-08364 Supplier Medchemexpress LLC Supplier No. HYW08738310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide 5-fluoride is a thalidomide-derived cereblon ligand used as a building block in PROTAC degraders and other bifunctional molecules. It is supplied as a high-purity solid reagent intended for research and chemical biology applications.

  • High purity solid (99.96%).
  • Molecular weight 276.22 g/mol.
  • Molecular formula C13H9FN2O4.
  • Available in laboratory pack sizes, including 10 G.
  • Storage: powder at -20°C for long-term, 4°C for shorter term; in solvent store at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.