Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-5-methyl | 40313-92-6 | 99.9% | 272.26 | 50 MG
SDP

Catalog No. 5000349529
Encompass_Preferred

Thalidomide-5-methyl is a cereblon (CRBN) ligand, based on Thalidomide, used in the recruitment of CRBN protein. It can be utilized for the synthesis of PROTACs and is intended for research use only.

  • Cereblon (CRBN) ligand
  • Recruits CRBN protein
  • Utilized for PROTAC synthesis
  • Molecular weight: 272.26
  • Solid appearance, gray to gray purple color
  • Soluble in DMSO (50 mg/mL with warming)
  • Recommended storage at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-49529 Supplier Medchemexpress LLC Supplier No. HY13954850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-5-methyl is a cereblon (CRBN) ligand, based on Thalidomide, used in the recruitment of CRBN protein. It can be utilized for the synthesis of PROTACs and is intended for research use only.

  • Cereblon (CRBN) ligand
  • Recruits CRBN protein
  • Utilized for PROTAC synthesis
  • Molecular weight: 272.26
  • Solid appearance, gray to gray purple color
  • Soluble in DMSO (50 mg/mL with warming)
  • Recommended storage at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.