Learn More
Medchemexpress LLC Thalidomide-5-methyl | 40313-92-6 | MFCD30290642 | 99.9% | 272.26 g·mol-1 | C14H12N2O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1395485MG
Encompass_Preferred
Thalidomide-5-methyl is a thalidomide-derived cereblon (CRBN) ligand used to recruit the CRBN E3 ligase subunit in chemical biology and targeted protein degradation applications. It is provided as a research-grade small molecule suitable for use as a CRBN-binding moiety in PROTAC synthesis.
  • Cereblon (CRBN) ligand for recruiting E3 ligase in PROTAC design.
  • High purity suitable for research applications (99.87%).
  • Molecular formula C14H12N2O4 and molecular weight 272.26 g·mol-1.
  • Supplied in milligram-scale quantities for synthesis and assay development.
  • Useful as a modular CRBN-binding building block in chemical biology experiments.
Catalog No. 50-002-23265