Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-5-NH-C-alkyne | 2357110-24-6 | MFCD34563706 | >98.0% | 311.29 g/mol | C16H13N3O4 | 100 MG
SDP

Catalog No. 5000284498
Encompass_Preferred

Thalidomide-5-NH-C-alkyne is an E3 ligase ligand-linker conjugate bearing a terminal alkyne for click-style conjugation, supplied for research use in small packs for assembling PROTACs and other bioconjugates.

  • Contains a terminal alkyne for click conjugation.
  • Functions as an E3 ligase ligand suitable for PROTAC assembly.
  • Purity >98.0%.
  • Molecular formula C16H13N3O4; molecular weight 311.29 g/mol.
  • CAS number 2357110-24-6 for substance identification.
  • Supplied as a 100 mg research pack.
  • For research use only; not for human or animal therapeutic use.

Catalog No. 50-002-84498 Supplier Medchemexpress LLC Supplier No. HYW674324100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-5-NH-C-alkyne is an E3 ligase ligand-linker conjugate bearing a terminal alkyne for click-style conjugation, supplied for research use in small packs for assembling PROTACs and other bioconjugates.

  • Contains a terminal alkyne for click conjugation.
  • Functions as an E3 ligase ligand suitable for PROTAC assembly.
  • Purity >98.0%.
  • Molecular formula C16H13N3O4; molecular weight 311.29 g/mol.
  • CAS number 2357110-24-6 for substance identification.
  • Supplied as a 100 mg research pack.
  • For research use only; not for human or animal therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.