Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-5-NH-C-alkyne | 2357110-24-6 | MFCD34563706 | 98.0% | 311.29 g/mol | C16H13N3O4 | 25 MG
SDP

Catalog No. 5000284949
Encompass_Preferred

Thalidomide-5-NH-C-alkyne is a thalidomide-derived E3 ligase ligand-linker conjugate bearing a terminal alkyne for click chemistry. It is supplied as a high-purity synthetic building block intended for conjugation in PROTAC synthesis and targeted protein degradation research.

  • Contains an E3 ligase ligand with a terminal alkyne for click chemistry.
  • Ready for conjugation to target molecules in PROTAC synthesis.
  • High purity: 98.0%.
  • Provided as 25 MG powder for small-scale research use.
  • Molecular weight 311.29 g/mol; formula C16H13N3O4.
  • Suitable for medicinal chemistry and chemical biology applications.

Catalog No. 50-002-84949 Supplier Medchemexpress LLC Supplier No. HYW67432425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-5-NH-C-alkyne is a thalidomide-derived E3 ligase ligand-linker conjugate bearing a terminal alkyne for click chemistry. It is supplied as a high-purity synthetic building block intended for conjugation in PROTAC synthesis and targeted protein degradation research.

  • Contains an E3 ligase ligand with a terminal alkyne for click chemistry.
  • Ready for conjugation to target molecules in PROTAC synthesis.
  • High purity: 98.0%.
  • Provided as 25 MG powder for small-scale research use.
  • Molecular weight 311.29 g/mol; formula C16H13N3O4.
  • Suitable for medicinal chemistry and chemical biology applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.