Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-5-NH2-C8-NH2 (TFA) | 2097509-49-2 | 514.49 g/mol | C23H29F3N4O6 | 10 MG
SDP

Catalog No. 5000285387
Encompass_Preferred

Thalidomide-5-NH2-C8-NH2 (TFA) is a thalidomide-derived intermediate provided as the trifluoroacetic acid salt for research use and compound synthesis. It is intended for laboratory applications only and should be handled using appropriate safety procedures; store sealed at -20°C away from moisture and light. In solvent, recommended storage is -80°C for up to 6 months or -20°C for up to 1 month.

  • Salt form: trifluoroacetic acid (TFA) salt.
  • Suitable for compound synthesis and as a building block.
  • Molecular formula: C23H29F3N4O6.
  • Molecular weight: 514.49 g/mol.
  • Storage: sealed at -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-85387 Supplier Medchemexpress LLC Supplier No. HY14887210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-5-NH2-C8-NH2 (TFA) is a thalidomide-derived intermediate provided as the trifluoroacetic acid salt for research use and compound synthesis. It is intended for laboratory applications only and should be handled using appropriate safety procedures; store sealed at -20°C away from moisture and light. In solvent, recommended storage is -80°C for up to 6 months or -20°C for up to 1 month.

  • Salt form: trifluoroacetic acid (TFA) salt.
  • Suitable for compound synthesis and as a building block.
  • Molecular formula: C23H29F3N4O6.
  • Molecular weight: 514.49 g/mol.
  • Storage: sealed at -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.