Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-5-nh2-c8-nh2 tfa | 2097509-49-2 | 98.3% | 514.49 | 100 MG
SDP

Catalog No. 5000361966
Encompass_Preferred

Thalidomide-5-NH2-C8-NH2 TFA is a chemical compound that can be used for compound synthesis. It is for research purposes only and not for sale to patients.

  • Appearance: Solid
  • Color: Light yellow to green yellow
  • Molecular Formula: C23H29F3N4O6

Catalog No. 50-003-61966 Supplier Medchemexpress LLC Supplier No. HY148872100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-5-NH2-C8-NH2 TFA is a chemical compound that can be used for compound synthesis. It is for research purposes only and not for sale to patients.

  • Appearance: Solid
  • Color: Light yellow to green yellow
  • Molecular Formula: C23H29F3N4O6

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.