Learn More
Medchemexpress LLC Thalidomide-5-NH2-CH2-COOH | 2412056-27-8 | 95.0% | C15H13N3O6 | 500 MG
SDP
Catalog No. 5000358222
Encompass_Preferred
Thalidomide-5-NH2-CH2-COOH (compound 114) is a potent and selective inhibitor of tropomyosin receptor kinase (Trk) and a ligand of E3 ligase. It has potential for researching various diseases, as extracted from patent WO2021170109A1.
  • Potent and selective Trk inhibitor
  • Ligand of E3 ligase
  • Researches various diseases including cancer, inflammation, and neurodegenerative conditions
  • Solid appearance
  • Color can range from light yellow to green yellow
Catalog No. 50-003-58222 Supplier Medchemexpress LLC Supplier No. HY46531500MG
Thalidomide-5-NH2-CH2-COOH (compound 114) is a potent and selective inhibitor of tropomyosin receptor kinase (Trk) and a ligand of E3 ligase. It has potential for researching various diseases, as extracted from patent WO2021170109A1.
  • Potent and selective Trk inhibitor
  • Ligand of E3 ligase
  • Researches various diseases including cancer, inflammation, and neurodegenerative conditions
  • Solid appearance
  • Color can range from light yellow to green yellow