Learn More
Medchemexpress LLC Thalidomide-5-NH2-CH2-COOH | 2412056-27-8 | 95.00% | C15H13N3O6 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY4653150MG
Encompass_Preferred
Thalidomide-5-NH2-CH2-COOH (compound 114) is a potent and selective inhibitor of tropomyosin receptor kinase (trk). It also functions as a ligand of E3 ligase and has potential for researching various diseases, as referenced in patent WO2021170109A1. This compound has a molecular weight of 331.28 and appears as a solid, ranging from light yellow to green yellow in color.
  • Potent and selective inhibitor of tropomyosin receptor kinase (trk)
  • Functions as a ligand of E3 ligase
  • Potential for researching various diseases
  • Appearance: solid, light yellow to green yellow
Catalog No. 50-003-59021