Learn More
Medchemexpress LLC Thalidomide-5-O-C10-NH2 hydrochloride | 99.4% | 465.97 g/mol | C23H32ClN3O5 | 10 MG
SDP
Catalog No. 5000287348
Encompass_Preferred
Thalidomide-5-O-C10-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand supplied as the hydrochloride salt for chemical biology research. It is used to recruit the CRBN E3 ubiquitin ligase in PROTAC and conjugate applications to enable targeted protein ubiquitination and degradation. Molecular formula C23H32ClN3O5; molecular weight 465.97 g/mol.
  • Used as a cereblon-binding ligand for PROTAC and conjugate design.
  • Supplied as the hydrochloride salt for improved handling and stability.
  • High purity suitable for research applications.
  • Available in small research pack sizes for synthesis and screening.
  • Provides a chemical handle designed for linker attachment to target ligands.
Catalog No. 50-002-87348 Supplier Medchemexpress LLC Supplier No. HY14956710MG
Thalidomide-5-O-C10-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand supplied as the hydrochloride salt for chemical biology research. It is used to recruit the CRBN E3 ubiquitin ligase in PROTAC and conjugate applications to enable targeted protein ubiquitination and degradation. Molecular formula C23H32ClN3O5; molecular weight 465.97 g/mol.
  • Used as a cereblon-binding ligand for PROTAC and conjugate design.
  • Supplied as the hydrochloride salt for improved handling and stability.
  • High purity suitable for research applications.
  • Available in small research pack sizes for synthesis and screening.
  • Provides a chemical handle designed for linker attachment to target ligands.