Learn More
Medchemexpress LLC Thalidomide-5-O-C11-NH2 hydrochloride | >98.0% | 480.00 | C24H34ClN3O5 | 10 MG
SDP
Catalog No. 5000287349
Encompass_Preferred
Thalidomide-5-O-C11-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand used to recruit the CRBN E3 ligase in targeted protein degradation constructs. Supplied as a white to off-white solid, it is intended for research use in PROTAC synthesis and ligand-linker conjugation. Handle under light protection and low-temperature storage to maintain stability.
  • Thalidomide-derived cereblon ligand for E3 ligase recruitment.
  • High purity (>98.0% HPLC) suitable for synthetic applications.
  • White to off-white solid, convenient for handling and weighing.
  • Stable under low-temperature, light-protected storage when handled under nitrogen.
  • Available in small research quantities for conjugation and PROTAC development.
Catalog No. 50-002-87349 Supplier Medchemexpress LLC Supplier No. HY14956810MG
Thalidomide-5-O-C11-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand used to recruit the CRBN E3 ligase in targeted protein degradation constructs. Supplied as a white to off-white solid, it is intended for research use in PROTAC synthesis and ligand-linker conjugation. Handle under light protection and low-temperature storage to maintain stability.
  • Thalidomide-derived cereblon ligand for E3 ligase recruitment.
  • High purity (>98.0% HPLC) suitable for synthetic applications.
  • White to off-white solid, convenient for handling and weighing.
  • Stable under low-temperature, light-protected storage when handled under nitrogen.
  • Available in small research quantities for conjugation and PROTAC development.