Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-5-O-C12-NH2 hydrochloride | 99.9% | 494.02 g/mol | C25H36ClN3O5 | 10 MG
SDP

Catalog No. 5000286618
Encompass_Preferred

Thalidomide-5-O-C12-NH2 hydrochloride is a thalidomide-derived cereblon ligand supplied as the hydrochloride salt for research applications. It is intended for use as an E3 ligase recruiter in PROTAC design and biochemical assays that engage CRBN. For research use only.

  • High purity 99.9%.
  • Molecular formula C25H36ClN3O5.
  • Molecular weight 494.02 g/mol.
  • Supplied in small milligram quantities, including 10 mg.
  • Suitable for ligand conjugation and targeted protein degradation studies.
  • Store protected from light; short-term at -20°C, long-term at -80°C in solvent.

Catalog No. 50-002-86618 Supplier Medchemexpress LLC Supplier No. HY14956910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-5-O-C12-NH2 hydrochloride is a thalidomide-derived cereblon ligand supplied as the hydrochloride salt for research applications. It is intended for use as an E3 ligase recruiter in PROTAC design and biochemical assays that engage CRBN. For research use only.

  • High purity 99.9%.
  • Molecular formula C25H36ClN3O5.
  • Molecular weight 494.02 g/mol.
  • Supplied in small milligram quantities, including 10 mg.
  • Suitable for ligand conjugation and targeted protein degradation studies.
  • Store protected from light; short-term at -20°C, long-term at -80°C in solvent.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.