Learn More
Medchemexpress LLC Thalidomide-5-O-C13-NH2 (hydrochloride) | 99.9% | 508.05 | 5 MG
SDP
Catalog No. 5000360831
Encompass_Preferred
Thalidomide-5-O-C13-NH2 hydrochloride is a cereblon ligand based on Thalidomide. It is utilized in the recruitment of CRBN protein and can be connected to a protein ligand by a linker to form PROTACs.
  • Cereblon ligand
  • Used in the recruitment of CRBN protein
  • Can be connected to a protein ligand by a linker
  • Forms PROTACs
Catalog No. 50-003-60831 Supplier Medchemexpress LLC Supplier No. HY1495705MG
Thalidomide-5-O-C13-NH2 hydrochloride is a cereblon ligand based on Thalidomide. It is utilized in the recruitment of CRBN protein and can be connected to a protein ligand by a linker to form PROTACs.
  • Cereblon ligand
  • Used in the recruitment of CRBN protein
  • Can be connected to a protein ligand by a linker
  • Forms PROTACs