Learn More
Medchemexpress LLC Thalidomide-5-O-C13-NH2 hydrochloride | 99.9% | 508.05 g/mol | C26H38ClN3O5 | 10 MG
SDP
Catalog No. 5000286811
Encompass_Preferred
Thalidomide-5-O-C13-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand supplied as the hydrochloride salt for research use in targeted protein degradation. It is intended for conjugation to target-binding ligands via linkers for PROTAC design and other bifunctional degrader constructs.
  • High purity suitable for research assays (approximately 99.9%).
  • Hydrochloride salt form for improved stability and handling.
  • Compatible with linker conjugation for PROTAC synthesis.
  • Recommended storage at -20°C; protect from light and oxygen.
  • Supplied in small milligram quantities for exploratory studies.
Catalog No. 50-002-86811 Supplier Medchemexpress LLC Supplier No. HY14957010MG
Thalidomide-5-O-C13-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand supplied as the hydrochloride salt for research use in targeted protein degradation. It is intended for conjugation to target-binding ligands via linkers for PROTAC design and other bifunctional degrader constructs.
  • High purity suitable for research assays (approximately 99.9%).
  • Hydrochloride salt form for improved stability and handling.
  • Compatible with linker conjugation for PROTAC synthesis.
  • Recommended storage at -20°C; protect from light and oxygen.
  • Supplied in small milligram quantities for exploratory studies.