Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-5-O-C14-NH2 hydrochloride | 97.7% | 522.08 g/mol | C27H40ClN3O5 | 10 MG
SDP

Catalog No. 5000286065
Encompass_Preferred

Thalidomide-5-O-C14-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand supplied for research use in targeted protein degradation and PROTAC development. It is provided as a small-quantity, high-purity solid suitable for conjugation through linkers to build bifunctional degraders and other CRBN-recruiting constructs.

  • High purity of 97.7%.
  • Molecular weight 522.08 g/mol and chemical formula C27H40ClN3O5.
  • White to off-white solid appearance.
  • Storage recommendations: -20 °C; in solvent: -80 °C (6 months) or -20 °C (1 month), protect from light, store under nitrogen.
  • Suitable for use as a CRBN-recruiting ligand in PROTAC synthesis.

Catalog No. 50-002-86065 Supplier Medchemexpress LLC Supplier No. HY14957110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-5-O-C14-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand supplied for research use in targeted protein degradation and PROTAC development. It is provided as a small-quantity, high-purity solid suitable for conjugation through linkers to build bifunctional degraders and other CRBN-recruiting constructs.

  • High purity of 97.7%.
  • Molecular weight 522.08 g/mol and chemical formula C27H40ClN3O5.
  • White to off-white solid appearance.
  • Storage recommendations: -20 °C; in solvent: -80 °C (6 months) or -20 °C (1 month), protect from light, store under nitrogen.
  • Suitable for use as a CRBN-recruiting ligand in PROTAC synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.