Learn More
Medchemexpress LLC Thalidomide-5-O-C2-NH2 hydrochloride | 2694727-89-2 | 98.0% | 353.76 g/mol | C15H16ClN3O5 | 10 MG
SDP
Catalog No. 5000285840
Encompass_Preferred
Thalidomide-5-O-C2-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand supplied as the hydrochloride salt. It is used as an E3 ligase ligand-linker conjugate in targeted protein degradation research and PROTAC synthesis, enabling the attachment of linkers or functional groups to the thalidomide scaffold for chemical biology applications.
  • High reported purity, 98.0%.
  • Supplied as the hydrochloride salt for improved handling.
  • Functions as a cereblon (CRBN) ligand for E3 ligase recruitment.
  • Suitable for PROTAC synthesis and ligand-linker conjugation.
  • Characterized by CAS 2694727-89-2 and molecular weight 353.76 g/mol.
  • Available in small research pack sizes such as 10 MG.
Catalog No. 50-002-85840 Supplier Medchemexpress LLC Supplier No. HY14955910MG
Thalidomide-5-O-C2-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand supplied as the hydrochloride salt. It is used as an E3 ligase ligand-linker conjugate in targeted protein degradation research and PROTAC synthesis, enabling the attachment of linkers or functional groups to the thalidomide scaffold for chemical biology applications.
  • High reported purity, 98.0%.
  • Supplied as the hydrochloride salt for improved handling.
  • Functions as a cereblon (CRBN) ligand for E3 ligase recruitment.
  • Suitable for PROTAC synthesis and ligand-linker conjugation.
  • Characterized by CAS 2694727-89-2 and molecular weight 353.76 g/mol.
  • Available in small research pack sizes such as 10 MG.