Learn More
Medchemexpress LLC Thalidomide-5-O-C2-NH2 hydrochloride | 2694727-89-2 | 99.6% | 353.76 | 5MG
SDP
Catalog No. 5000360999
Encompass_Preferred
Thalidomide-5-O-C2-NH2 hydrochloride is a cereblon ligand based on Thalidomide, designed for the recruitment of CRBN protein. This compound can be linked to a protein ligand via a linker to form PROTACs, which are relevant in cancer targeted therapy and metabolism research.
  • Used for the recruitment of CRBN protein
  • Can be connected to a protein ligand by a linker to form PROTACs
  • Target is Cereblon
  • Involved in E3 ligase ligand-linker conjugates, apoptosis, and autophagy research
Catalog No. 50-003-60999 Supplier Medchemexpress LLC Supplier No. HY1495595MG
Thalidomide-5-O-C2-NH2 hydrochloride is a cereblon ligand based on Thalidomide, designed for the recruitment of CRBN protein. This compound can be linked to a protein ligand via a linker to form PROTACs, which are relevant in cancer targeted therapy and metabolism research.
  • Used for the recruitment of CRBN protein
  • Can be connected to a protein ligand by a linker to form PROTACs
  • Target is Cereblon
  • Involved in E3 ligase ligand-linker conjugates, apoptosis, and autophagy research