Learn More
Medchemexpress LLC Thalidomide-5-O-C3-NH2 hydrochloride | 2694727-94-9 | MFCD34475114 | 99.1% | 367.78 g/mol | C16H18ClN3O5 | 10 MG
SDP
Catalog No. 5000286617
Encompass_Preferred
Thalidomide-5-O-C3-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand supplied as the hydrochloride salt for use as a building block in targeted protein degradation research and PROTAC design. It is provided as a solid with supporting analytical data for research use.
  • Acts as a cereblon-binding ligand for CRBN recruitment.
  • Enables linker conjugation for PROTAC assembly.
  • Supplied with high purity (99.08%).
  • Provided as a solid suitable for long-term storage under recommended conditions.
  • Analytical data and COA available to support research use.
Catalog No. 50-002-86617 Supplier Medchemexpress LLC Supplier No. HY14956010MG
Thalidomide-5-O-C3-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand supplied as the hydrochloride salt for use as a building block in targeted protein degradation research and PROTAC design. It is provided as a solid with supporting analytical data for research use.
  • Acts as a cereblon-binding ligand for CRBN recruitment.
  • Enables linker conjugation for PROTAC assembly.
  • Supplied with high purity (99.08%).
  • Provided as a solid suitable for long-term storage under recommended conditions.
  • Analytical data and COA available to support research use.