Learn More
Medchemexpress LLC Thalidomide-5-O-C4-NH2 hydrochloride | 2694727-93-8 | 100.0% | 381.81 g/mol | C17H20ClN3O5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY14956110MG
Encompass_Preferred
Thalidomide-5-O-C4-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand intended for chemical biology research and PROTAC design. Supplied as a white to off-white solid in small milligram quantities, the compound functions as an E3 ligase recruiting moiety that can be connected to target ligands via linkers for targeted protein degradation studies.
  • High purity suitable for research applications (reported ~99.98%).
  • Acts as a cereblon-binding ligand for E3 ligase recruitment.
  • Designed for PROTAC synthesis and linker conjugation.
  • Available in small milligram pack sizes for screening and synthesis.
  • Solid form, white to off-white, stable under recommended storage.
  • Includes CAS number 2694727-93-8 for unambiguous identification.
Catalog No. 50-002-85842