Learn More
Medchemexpress LLC Thalidomide-5-O-C5-NH2 hydrochloride | 2761385-94-6 | 395.84 | C18H22ClN3O5 | 10 MG
SDP
Catalog No. 5000286616
Encompass_Preferred
Thalidomide-5-O-C5-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand provided as the hydrochloride salt for use in chemical biology research. It is designed for incorporation into ligand-linker conjugates and PROTAC constructs to recruit CRBN and enable targeted protein degradation. The material is supplied as a research-grade powder with recommended cold, light-protected storage conditions.
Catalog No. 50-002-86616 Supplier Medchemexpress LLC Supplier No. HY14956210MG
Thalidomide-5-O-C5-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand provided as the hydrochloride salt for use in chemical biology research. It is designed for incorporation into ligand-linker conjugates and PROTAC constructs to recruit CRBN and enable targeted protein degradation. The material is supplied as a research-grade powder with recommended cold, light-protected storage conditions.