Learn More
Medchemexpress LLC Thalidomide-5-O-C6-NH2 hydrochloride | 2761398-05-2 | 409.86 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1495635MG
Encompass_Preferred
Thalidomide-5-O-C6-NH2 hydrochloride is a cereblon ligand based on Thalidomide. It is utilized in the recruitment of CRBN protein and can be linked to a ligand for protein via a linker to form PROTACs. It is a solid with a white to off-white appearance.
  • Cereblon ligand
  • Utilized in CRBN protein recruitment
  • Can form PROTACs when linked to a protein ligand
  • Available as a white to off-white solid
  • Shipped at room temperature
  • Stored at -20°C, protected from light and under nitrogen
Catalog No. 50-003-60272