Learn More
Medchemexpress LLC Thalidomide-5-O-C6-NH2 hydrochloride | 2761398-05-2 | 409.86 | C19H24ClN3O5 | 10 MG
SDP
Catalog No. 5000287347
Encompass_Preferred
Thalidomide-5-O-C6-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand used as an E3 ligase recruiting moiety in PROTAC development and chemical biology research. The compound contains a six-carbon linker terminating in a primary amine and is supplied as the hydrochloride salt to aid handling and stability for research applications.
  • Used as a cereblon ligand to recruit CRBN in PROTAC and degrader design.
  • Contains a C6 linker with a terminal amine for conjugation to linkers or target ligands.
  • Provided as a hydrochloride salt for improved handling and solubility.
  • Solid form suitable for storage under recommended conditions.
  • Intended for research use; consult vendor documentation or COA for purity and analytical data.
Catalog No. 50-002-87347 Supplier Medchemexpress LLC Supplier No. HY14956310MG
Thalidomide-5-O-C6-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand used as an E3 ligase recruiting moiety in PROTAC development and chemical biology research. The compound contains a six-carbon linker terminating in a primary amine and is supplied as the hydrochloride salt to aid handling and stability for research applications.
  • Used as a cereblon ligand to recruit CRBN in PROTAC and degrader design.
  • Contains a C6 linker with a terminal amine for conjugation to linkers or target ligands.
  • Provided as a hydrochloride salt for improved handling and solubility.
  • Solid form suitable for storage under recommended conditions.
  • Intended for research use; consult vendor documentation or COA for purity and analytical data.