Learn More
Medchemexpress LLC Thalidomide-5-O-C7-NH2 hydrochloride | 98.1% | 423.89 g/mol | C20H26ClN3O5 | 10 MG
SDP
Catalog No. 5000286810
Encompass_Preferred
Thalidomide-5-O-C7-NH2 hydrochloride is a thalidomide-derived cereblon ligand supplied as the hydrochloride salt for use in chemical biology and targeted protein degradation research. It is designed to be connected to target ligands via a linker to build PROTACs and other CRBN-recruiting constructs, and includes storage recommendations to preserve compound integrity.
  • Facilitates cereblon (CRBN) recruitment for targeted degradation studies.
  • Hydrochloride salt form for improved handling and stability.
  • Compatible with linker conjugation to form PROTAC molecules.
  • Provided with documented molecular and storage data to support experimental planning.
  • Suitable for research applications requiring characterized E3 ligase ligands.
Catalog No. 50-002-86810 Supplier Medchemexpress LLC Supplier No. HY14956410MG
Thalidomide-5-O-C7-NH2 hydrochloride is a thalidomide-derived cereblon ligand supplied as the hydrochloride salt for use in chemical biology and targeted protein degradation research. It is designed to be connected to target ligands via a linker to build PROTACs and other CRBN-recruiting constructs, and includes storage recommendations to preserve compound integrity.
  • Facilitates cereblon (CRBN) recruitment for targeted degradation studies.
  • Hydrochloride salt form for improved handling and stability.
  • Compatible with linker conjugation to form PROTAC molecules.
  • Provided with documented molecular and storage data to support experimental planning.
  • Suitable for research applications requiring characterized E3 ligase ligands.