Learn More
Medchemexpress LLC Thalidomide-5-O-C7-NH2 hydrochloride | 98.13% | 423.89 | 5 MG
SDP
Catalog No. 5000360321
Encompass_Preferred
Thalidomide-5-O-C7-NH2 hydrochloride is a Thalidomide (HY-10984)-based cereblon ligand that is used for recruiting CRBN protein. It can be connected to a ligand for protein by a linker to form PROTACs.
  • Used in the recruitment of CRBN protein
  • Can be connected to a ligand for protein by a linker
  • Forms PROTACs
  • For research use only
Catalog No. 50-003-60321 Supplier Medchemexpress LLC Supplier No. HY1495645MG
Thalidomide-5-O-C7-NH2 hydrochloride is a Thalidomide (HY-10984)-based cereblon ligand that is used for recruiting CRBN protein. It can be connected to a ligand for protein by a linker to form PROTACs.
  • Used in the recruitment of CRBN protein
  • Can be connected to a ligand for protein by a linker
  • Forms PROTACs
  • For research use only