Learn More
Medchemexpress LLC Thalidomide-5-O-C8-NH2 hydrochloride | 2636798-38-2 | >95.0% | 437.92 g/mol | C21H28ClN3O5 | 10 MG
SDP
Catalog No. 5000286656
Encompass_Preferred
Thalidomide-5-O-C8-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand supplied as the hydrochloride salt for research applications. It is used as a recruiting moiety in PROTACs and other E3 ligase-targeting conjugates to facilitate targeted protein degradation and chemical biology studies. Supplied as a white to off-white solid in small laboratory pack sizes, it supports synthetic conjugation through an alkyl linker.
  • Serves as a cereblon-recruiting ligand for targeted protein degradation
  • Supplied as a hydrochloride salt for improved stability and handling
  • Solid form suitable for storage and synthetic manipulation
  • Provides an alkyl linker attachment point for PROTAC assembly
  • Compatible with standard organic synthesis and conjugation workflows
  • Intended for research use in chemical biology and drug discovery
Catalog No. 50-002-86656 Supplier Medchemexpress LLC Supplier No. HY14956510MG
Thalidomide-5-O-C8-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand supplied as the hydrochloride salt for research applications. It is used as a recruiting moiety in PROTACs and other E3 ligase-targeting conjugates to facilitate targeted protein degradation and chemical biology studies. Supplied as a white to off-white solid in small laboratory pack sizes, it supports synthetic conjugation through an alkyl linker.
  • Serves as a cereblon-recruiting ligand for targeted protein degradation
  • Supplied as a hydrochloride salt for improved stability and handling
  • Solid form suitable for storage and synthetic manipulation
  • Provides an alkyl linker attachment point for PROTAC assembly
  • Compatible with standard organic synthesis and conjugation workflows
  • Intended for research use in chemical biology and drug discovery