Learn More
Medchemexpress LLC Thalidomide-5-O-C8-NH2 hydrochloride | ≥98% | 437.92 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1495655MG
Encompass_Preferred
Thalidomide-5-O-C8-NH2 hydrochloride is a cereblon ligand derived from Thalidomide. It is utilized to recruit CRBN protein and can be linked to a target protein ligand via a linker to form PROTACs.
  • It is a cereblon ligand.
  • It is used in the recruitment of CRBN protein.
  • It can be connected to a ligand for protein by a linker to form PROTACs.
  • It is intended for research purposes only and not for sale to patients.
Catalog No. 50-003-59460