Learn More
Medchemexpress LLC Thalidomide-5-o-c9-nh2 hydrochloride | 99.6% | 451.94 g·mol⁻1 | C22H30ClN3O5 | 10 MG
SDP
Catalog No. 5000285776
Encompass_Preferred
Thalidomide-5-O-C9-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand intended for chemical biology research. Supplied as the hydrochloride salt, it is used as an E3 ligase recruiting moiety in PROTACs and related conjugates. The product is reported with molecular formula C22H30ClN3O5 and molecular weight 451.94 g·mol⁻1.
  • High purity: 99.6%.
  • Hydrochloride salt form for improved handling and solubility.
  • Designed for recruitment of cereblon (CRBN) in PROTAC construction.
  • Suitable for synthesis, assay development, and linker conjugation.
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-85776 Supplier Medchemexpress LLC Supplier No. HY14956610MG
Thalidomide-5-O-C9-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand intended for chemical biology research. Supplied as the hydrochloride salt, it is used as an E3 ligase recruiting moiety in PROTACs and related conjugates. The product is reported with molecular formula C22H30ClN3O5 and molecular weight 451.94 g·mol⁻1.
  • High purity: 99.6%.
  • Hydrochloride salt form for improved handling and solubility.
  • Designed for recruitment of cereblon (CRBN) in PROTAC construction.
  • Suitable for synthesis, assay development, and linker conjugation.
  • Available in small research pack sizes for laboratory use.