Learn More
Medchemexpress LLC Thalidomide-5-O-CH2-COOH | 2467425-53-0 | 95.0% | 332.26 g/mol | C15H12N2O7 | 25 MG
SDP
Catalog No. 5000284631
Encompass_Preferred
Thalidomide-5-O-CH2-COOH is a thalidomide-derived E3 ligase ligand-linker conjugate used as a cereblon ligand in chemical biology and targeted protein degradation research. It is supplied as a solid reagent intended for in vitro studies and synthetic applications.
  • Acts as a cereblon (CRBN) ligand for targeted protein degradation studies.
  • Designed as a ligand-linker conjugate to enable attachment to target-binding warheads.
  • High solubility in DMSO (125 mg/mL) facilitates preparation of concentrated stocks.
  • Supplied with typical purity of 95.0% suitable for research use.
  • Molecular weight 332.26 g/mol and formula C15H12N2O7 for accurate dosing.
  • Stable as a powder when stored frozen for long-term retention.
  • Accompanied by datasheet, certificate of analysis, and safety data for handling guidance.
Catalog No. 50-002-84631 Supplier Medchemexpress LLC Supplier No. HYW58451825MG
Thalidomide-5-O-CH2-COOH is a thalidomide-derived E3 ligase ligand-linker conjugate used as a cereblon ligand in chemical biology and targeted protein degradation research. It is supplied as a solid reagent intended for in vitro studies and synthetic applications.
  • Acts as a cereblon (CRBN) ligand for targeted protein degradation studies.
  • Designed as a ligand-linker conjugate to enable attachment to target-binding warheads.
  • High solubility in DMSO (125 mg/mL) facilitates preparation of concentrated stocks.
  • Supplied with typical purity of 95.0% suitable for research use.
  • Molecular weight 332.26 g/mol and formula C15H12N2O7 for accurate dosing.
  • Stable as a powder when stored frozen for long-term retention.
  • Accompanied by datasheet, certificate of analysis, and safety data for handling guidance.