Learn More
Medchemexpress LLC Thalidomide-5-O-CH2-COOH | 2467425-53-0 | MFCD34186095 | 95.0% | 332.26 | C15H12N2O7 | 5 MG
SDP
Catalog No. 5000284271
Encompass_Preferred
Thalidomide-5-O-CH2-COOH is an E3 ligase ligand-linker conjugate used as a cereblon (CRBN) ligand and an intermediate in PROTAC and conjugate synthesis. The compound is fully characterized (CAS 2467425-53-0, C15H12N2O7, MW 332.26) and is provided in small milligram quantities for research use with supporting analytical documentation.
  • Functions as a cereblon (CRBN) ligand for E3 ligase recruitment.
  • Serves as an intermediate for PROTAC and ligand-linker conjugation.
  • Characterized by CAS 2467425-53-0 and formula C15H12N2O7.
  • Supplied with datasheet, certificate of analysis, SDS, and spectral data.
  • Typical purity approximately 95.0% by HPLC.
  • Available in milligram pack sizes suitable for medicinal chemistry research.
Catalog No. 50-002-84271 Supplier Medchemexpress LLC Supplier No. HYW5845185MG
Thalidomide-5-O-CH2-COOH is an E3 ligase ligand-linker conjugate used as a cereblon (CRBN) ligand and an intermediate in PROTAC and conjugate synthesis. The compound is fully characterized (CAS 2467425-53-0, C15H12N2O7, MW 332.26) and is provided in small milligram quantities for research use with supporting analytical documentation.
  • Functions as a cereblon (CRBN) ligand for E3 ligase recruitment.
  • Serves as an intermediate for PROTAC and ligand-linker conjugation.
  • Characterized by CAS 2467425-53-0 and formula C15H12N2O7.
  • Supplied with datasheet, certificate of analysis, SDS, and spectral data.
  • Typical purity approximately 95.0% by HPLC.
  • Available in milligram pack sizes suitable for medicinal chemistry research.