Learn More
Medchemexpress LLC Thalidomide-5-O-CH2-COOH | 2467425-53-0 | MFCD34186095 | 95.0% | 332.26 g/mol | C15H12N2O7 | 50 MG
SDP
Catalog No. 5000283864
Encompass_Preferred
Thalidomide-5-O-CH2-COOH is an E3 ligase ligand-linker conjugate used as a cereblon (CRBN) ligand and an intermediate for the synthesis of PROTAC molecules. Supplied as a solid with supplier-characterized purity, it is intended for research applications in targeted protein degradation.
  • Acts as a cereblon ligand for targeted protein degradation research.
  • Suitable intermediate for PROTAC synthesis and conjugation chemistry.
  • Approximately 95.0% purity, characterized by supplier analytical data.
  • Solid form, light yellow to brown appearance for easy handling.
  • Good solubility in DMSO for in vitro formulation and assays.
  • Stable when stored under recommended conditions for long-term use.
Catalog No. 50-002-83864 Supplier Medchemexpress LLC Supplier No. HYW58451850MG
Thalidomide-5-O-CH2-COOH is an E3 ligase ligand-linker conjugate used as a cereblon (CRBN) ligand and an intermediate for the synthesis of PROTAC molecules. Supplied as a solid with supplier-characterized purity, it is intended for research applications in targeted protein degradation.
  • Acts as a cereblon ligand for targeted protein degradation research.
  • Suitable intermediate for PROTAC synthesis and conjugation chemistry.
  • Approximately 95.0% purity, characterized by supplier analytical data.
  • Solid form, light yellow to brown appearance for easy handling.
  • Good solubility in DMSO for in vitro formulation and assays.
  • Stable when stored under recommended conditions for long-term use.