Learn More
Medchemexpress LLC Thalidomide-5-OH | 64567-60-8 | 98.2% | 274.23 | 25 G
SDP
Catalog No. 5000402679
Encompass_Preferred
Thalidomide-5-OH is a thalidomide-based cereblon ligand. It is used in the recruitment of CRBN protein and can be connected to a target protein ligand via a linker to form PROTACs.
  • Thalidomide-based cereblon ligand
  • Recruits CRBN protein
  • Connects to protein ligands via a linker to form PROTACs
  • Targets cereblon
Catalog No. 50-004-02679 Supplier Medchemexpress LLC Supplier No. 25GHY23095
Thalidomide-5-OH is a thalidomide-based cereblon ligand. It is used in the recruitment of CRBN protein and can be connected to a target protein ligand via a linker to form PROTACs.
  • Thalidomide-based cereblon ligand
  • Recruits CRBN protein
  • Connects to protein ligands via a linker to form PROTACs
  • Targets cereblon