Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-5-OH | 64567-60-8 | 98.23% | 274.23 | 5 G
SDP

Catalog No. 5000400451
Encompass_Preferred

Thalidomide-5-OH is a thalidomide-based cereblon ligand used in the recruitment of CRBN protein. It can be linked to a target protein ligand via a linker to create PROTACs. This product is intended for research use only.

  • Utilized in the recruitment of CRBN protein
  • Can be linked to a ligand for protein by a linker to form PROTACs
  • For research use only
  • Target cereblon
  • Solid appearance
  • Color ranges from gray to brown
  • Soluble in DMSO at 62.5 mg/mL

Catalog No. 50-004-00451 Supplier Medchemexpress LLC Supplier No. HY230955G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-5-OH is a thalidomide-based cereblon ligand used in the recruitment of CRBN protein. It can be linked to a target protein ligand via a linker to create PROTACs. This product is intended for research use only.

  • Utilized in the recruitment of CRBN protein
  • Can be linked to a ligand for protein by a linker to form PROTACs
  • For research use only
  • Target cereblon
  • Solid appearance
  • Color ranges from gray to brown
  • Soluble in DMSO at 62.5 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.