Learn More
Medchemexpress LLC Thalidomide-5-PEG2-Cl | 2230956-57-5 | 96.6% | 380.78 | 250 MG
SDP
Catalog No. 5000348856
Encompass_Preferred
Thalidomide-5-PEG2-Cl is a Thalidomide-based cereblon ligand that plays a crucial role in recruiting CRBN protein. It can be utilized to form PROTACs by linking to a target protein ligand. PROTACs are designed to selectively degrade target proteins within the intracellular ubiquitin-proteasome system by exploiting E3 ubiquitin ligase and target protein interactions.
  • Thalidomide-based cereblon ligand
  • Recruits CRBN protein
  • Can be linked to form PROTACs
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-48856 Supplier Medchemexpress LLC Supplier No. 250MGHY139543
Thalidomide-5-PEG2-Cl is a Thalidomide-based cereblon ligand that plays a crucial role in recruiting CRBN protein. It can be utilized to form PROTACs by linking to a target protein ligand. PROTACs are designed to selectively degrade target proteins within the intracellular ubiquitin-proteasome system by exploiting E3 ubiquitin ligase and target protein interactions.
  • Thalidomide-based cereblon ligand
  • Recruits CRBN protein
  • Can be linked to form PROTACs
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins