Learn More
Medchemexpress LLC Thalidomide-5-peg2-cl | 2230956-57-5 | 96.6% | 380.78 g/mol | C17H17ClN2O6 | 5 MG
SDP
Catalog No. 5000223125
Encompass_Preferred
Thalidomide-5-PEG2-Cl is a thalidomide-derived cereblon ligand intended for use as the E3 ligase-recruiting component in proteolysis-targeting chimera (PROTAC) synthesis. It contains a two-unit polyethylene glycol linker and a terminal chloro functional group to facilitate conjugation to target ligands. Intended for research use only.
  • Enables cereblon recruitment in PROTAC constructs.
  • Contains a PEG2 linker for flexible conjugation.
  • Terminal chloro group for facile chemical coupling.
  • High purity suitable for research applications.
  • Available in small milligram quantities for screening and synthesis.
Catalog No. 50-002-23125 Supplier Medchemexpress LLC Supplier No. HY1395435MG
Thalidomide-5-PEG2-Cl is a thalidomide-derived cereblon ligand intended for use as the E3 ligase-recruiting component in proteolysis-targeting chimera (PROTAC) synthesis. It contains a two-unit polyethylene glycol linker and a terminal chloro functional group to facilitate conjugation to target ligands. Intended for research use only.
  • Enables cereblon recruitment in PROTAC constructs.
  • Contains a PEG2 linker for flexible conjugation.
  • Terminal chloro group for facile chemical coupling.
  • High purity suitable for research applications.
  • Available in small milligram quantities for screening and synthesis.