Learn More
Medchemexpress LLC Thalidomide-5-PEG3-NH2 hydrochloride | 2761385-81-1 | 97% | 441.86 | 1 MG
SDP
Catalog No. 5000351978
Encompass_Preferred
Thalidomide-5-PEG3-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology. It has an IC 50 of Cereblon.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Thalidomide-based cereblon ligand
  • Used in PROTAC technology
  • Solid form
  • White to off-white color
Catalog No. 50-003-51978 Supplier Medchemexpress LLC Supplier No. HY138786A1MG
Thalidomide-5-PEG3-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology. It has an IC 50 of Cereblon.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Thalidomide-based cereblon ligand
  • Used in PROTAC technology
  • Solid form
  • White to off-white color