Learn More
Medchemexpress LLC Thalidomide-5-PEG3-NH2 hydrochloride | 2761385-81-1 | 97.9% | 441.86 g·mol⁻1 | C19H24ClN3O7 | 5 MG
SDP
Catalog No. 5000221997
Encompass_Preferred
Thalidomide-5-PEG3-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that combines a thalidomide-based cereblon ligand with a PEG3 linker terminating in a primary amine. Supplied as the hydrochloride salt, it is intended as a building block for PROTAC design and bioconjugation chemistry.
  • Contains a PEG3 linker with a terminal primary amine for conjugation.
  • Supplied as hydrochloride salt to aid handling and stability.
  • Intended for use as an E3 ligase recruiting ligand in PROTAC synthesis.
  • High reported purity suitable for research applications.
  • Available in small research sizes for medicinal chemistry workflows.
Catalog No. 50-002-21997 Supplier Medchemexpress LLC Supplier No. HY138786A5MG
Thalidomide-5-PEG3-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that combines a thalidomide-based cereblon ligand with a PEG3 linker terminating in a primary amine. Supplied as the hydrochloride salt, it is intended as a building block for PROTAC design and bioconjugation chemistry.
  • Contains a PEG3 linker with a terminal primary amine for conjugation.
  • Supplied as hydrochloride salt to aid handling and stability.
  • Intended for use as an E3 ligase recruiting ligand in PROTAC synthesis.
  • High reported purity suitable for research applications.
  • Available in small research sizes for medicinal chemistry workflows.