Learn More
Medchemexpress LLC Thalidomide-5-PEG3-NH2 hydrochloride | 2761385-81-1 | MFCD35061671; MFCD32068377 | 97.9% | C19H24ClN3O7 | 10MG
SDP
Supplier:  Medchemexpress LLC HY138786A10MG
Encompass_Preferred
Thalidomide-5-PEG3-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) E3 ligase ligand-linker conjugate supplied as the hydrochloride salt. It is intended for research use in PROTAC development and E3 ligase recruitment studies, providing a PEG3 linker terminating in a primary amine for convenient conjugation to target ligands.
  • Hydrochloride salt for improved stability and handling.
  • PEG3 linker with terminal primary amine for conjugation.
  • Cereblon (CRBN) ligand suitable for E3 ligase recruitment in PROTACs.
  • High purity appropriate for research synthesis and assays.
  • Available in small pack sizes for synthetic workflows.
Catalog No. 50-002-60640