Learn More
Medchemexpress LLC Thalidomide-5-peg4-nh2 | 2743434-16-2 | 95.21% | 449.45 | 100 MG
SDP
Catalog No. 5000349582
Encompass_Preferred
This compound is an E3 ligase ligand-linker conjugate that uses a Thalidomide-based cereblon ligand and a PROTAC linker. It is intended for research use only, with a recommendation to use its more stable hydrochloride salt form due to the free compound's instability.
  • Designed for PROTAC technology.
  • Incorporates a cereblon ligand.
  • Contains a versatile linker.
  • Suitable for research applications.
  • Available in a more stable salt form.
Catalog No. 50-003-49582 Supplier Medchemexpress LLC Supplier No. HY138785A100MG
This compound is an E3 ligase ligand-linker conjugate that uses a Thalidomide-based cereblon ligand and a PROTAC linker. It is intended for research use only, with a recommendation to use its more stable hydrochloride salt form due to the free compound's instability.
  • Designed for PROTAC technology.
  • Incorporates a cereblon ligand.
  • Contains a versatile linker.
  • Suitable for research applications.
  • Available in a more stable salt form.