Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-5-PEG4-NH2 hydrochloride | 2743434-24-2 | 95.2% | 485.92 g/mol | C21H28ClN3O8 | 5 MG
SDP

Catalog No. 5000223428
Encompass_Preferred

Thalidomide-5-PEG4-NH2 hydrochloride is a thalidomide-derived E3 ligase ligand-linker conjugate that combines a cereblon-binding thalidomide moiety with a four-unit PEG linker. It is intended as a research reagent for PROTAC design and targeted protein degradation studies and is supplied as a light yellow to yellow oil with reported high purity.

  • Designed as an E3 ligase ligand-linker conjugate for PROTAC synthesis.
  • Contains a cereblon-binding thalidomide moiety with a PEG4 linker for linker flexibility.
  • High reported purity suitable for research applications (95.2%).
  • Soluble in DMSO at 100 mg/mL; ultrasonic agitation recommended.
  • Light yellow to yellow oil; easy to handle in synthetic workflows.
  • Store sealed away from moisture; recommended storage at -20°C (in solvent: -80°C for long term).

Catalog No. 50-002-23428 Supplier Medchemexpress LLC Supplier No. HY138785A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-5-PEG4-NH2 hydrochloride is a thalidomide-derived E3 ligase ligand-linker conjugate that combines a cereblon-binding thalidomide moiety with a four-unit PEG linker. It is intended as a research reagent for PROTAC design and targeted protein degradation studies and is supplied as a light yellow to yellow oil with reported high purity.

  • Designed as an E3 ligase ligand-linker conjugate for PROTAC synthesis.
  • Contains a cereblon-binding thalidomide moiety with a PEG4 linker for linker flexibility.
  • High reported purity suitable for research applications (95.2%).
  • Soluble in DMSO at 100 mg/mL; ultrasonic agitation recommended.
  • Light yellow to yellow oil; easy to handle in synthetic workflows.
  • Store sealed away from moisture; recommended storage at -20°C (in solvent: -80°C for long term).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.