Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-5-PEG4-NH2 hydrochloride | 2743434-24-2 | 95.2% | C21H28ClN3O8 | 10MG
SDP

Catalog No. 5000261958
Encompass_Preferred

Thalidomide-5-PEG4-NH2 hydrochloride is a thalidomide-derived cereblon ligand conjugated to a PEG4 linker and supplied as the hydrochloride salt. It serves as an E3 ligase ligand-linker building block for PROTAC design and related chemical biology research.

  • Incorporates thalidomide-based cereblon ligand for E3 ligase recruitment.
  • PEG4 linker provides a flexible spacer for bifunctional molecule design.
  • Hydrochloride salt form improves handling and solubility properties.
  • Intended for use in proteolysis-targeting chimera (PROTAC) synthesis.
  • Molecular weight 485.92 g/mol; chemical formula C21H28ClN3O8.
  • Manufacturer-reported purity 95.2% suitable for research applications.

Catalog No. 50-002-61958 Supplier Medchemexpress LLC Supplier No. HY138785A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-5-PEG4-NH2 hydrochloride is a thalidomide-derived cereblon ligand conjugated to a PEG4 linker and supplied as the hydrochloride salt. It serves as an E3 ligase ligand-linker building block for PROTAC design and related chemical biology research.

  • Incorporates thalidomide-based cereblon ligand for E3 ligase recruitment.
  • PEG4 linker provides a flexible spacer for bifunctional molecule design.
  • Hydrochloride salt form improves handling and solubility properties.
  • Intended for use in proteolysis-targeting chimera (PROTAC) synthesis.
  • Molecular weight 485.92 g/mol; chemical formula C21H28ClN3O8.
  • Manufacturer-reported purity 95.2% suitable for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.