Learn More
Medchemexpress LLC Thalidomide-5-PEG7-NH2 hydrochloride | 618.07 g/mol | C27H40ClN3O11 | 10 MG

Supplier: Medchemexpress LLC HY15525710MG
Thalidomide-5-PEG7-NH2 hydrochloride is a thalidomide-derived cereblon ligand with a PEG7 spacer provided as the hydrochloride salt. It is designed for use as an E3 ligase ligand in PROTAC and ligand-linker conjugate synthesis and is supplied as a solid for research applications.
- Thalidomide-derived cereblon ligand for recruitment of CRBN.
- PEG7 spacer offers a flexible linker for conjugation to target ligands.
- Provided as the hydrochloride salt to aid handling and solubility.
- White to off-white solid physical form.
- Molecular formula C27H40ClN3O11 and molecular weight 618.07 g/mol.
- Store sealed at -20°C; in solution store at -80°C for up to 6 months or at -20°C for up to 1 month.
- Available in small mg quantities for laboratory research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.