Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-5-propargyl | 2226303-74-6 | 95.9% | 312.28 g/mol | C16H12N2O5 | 100 MG
SDP

Catalog No. 5000280967
Encompass_Preferred

Thalidomide-5-propargyl is a propargyl-modified thalidomide derivative used as a cereblon ligand in chemical biology and PROTAC research. It provides an alkyne handle for linker attachment and click chemistry, and is supplied as a solid for research use only.

  • Acts as a cereblon ligand to recruit CRBN protein.
  • Contains a propargyl (alkynyl) handle for triazole formation via click chemistry.
  • Suitable as a building block for PROTAC synthesis and ligand conjugation.
  • Offered at high purity (~95.9%) for research applications.
  • Recommended storage: 4°C; in solution under nitrogen at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-80967 Supplier Medchemexpress LLC Supplier No. HYW457950100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-5-propargyl is a propargyl-modified thalidomide derivative used as a cereblon ligand in chemical biology and PROTAC research. It provides an alkyne handle for linker attachment and click chemistry, and is supplied as a solid for research use only.

  • Acts as a cereblon ligand to recruit CRBN protein.
  • Contains a propargyl (alkynyl) handle for triazole formation via click chemistry.
  • Suitable as a building block for PROTAC synthesis and ligand conjugation.
  • Offered at high purity (~95.9%) for research applications.
  • Recommended storage: 4°C; in solution under nitrogen at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.