Learn More
Medchemexpress LLC Thalidomide-5-propargyl | 2226303-74-6 | 95.9% | 312.28 g/mol | C16H12N2O5 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW45795025MG
Encompass_Preferred
Thalidomide-5-propargyl is a propargyl-modified thalidomide derivative used as a cereblon ligand in chemical biology and PROTAC development. Supplied as a white to off-white solid for research use only, it is characterized by reported purity, CAS identifier, molecular formula, and recommended storage conditions; product documentation includes a data sheet and SDS for handling and safety guidance.
  • Acts as a cereblon ligand for recruitment of CRBN in targeted protein degradation studies.
  • Contains a propargyl handle enabling bioorthogonal conjugation and linker attachment.
  • High reported purity suitable for research applications.
  • White to off-white solid with recommended storage at 4°C under nitrogen.
  • Documentation includes data sheet and SDS for safe handling.
Catalog No. 50-002-81107