Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-5-propargyl | 2226303-74-6 | 95.9% | 312.28 g·mol⁻¹ | C16H12N2O5 | 50 MG
SDP

Supplier:  Medchemexpress LLC HYW45795050MG

Encompass_Preferred

Thalidomide-5-propargyl is a propargyl-modified thalidomide derivative that acts as a cereblon (CRBN) ligand for use in targeted protein degradation research. The terminal alkyne enables click chemistry and triazole formation, making the compound a versatile intermediate for PROTAC synthesis and medicinal chemistry applications.

  • Functions as a cereblon ligand for CRBN recruitment.
  • Contains a terminal alkyne suitable for click chemistry.
  • Suitable as an intermediate in PROTAC synthesis.
  • Provided with high purity (95.87%).
  • Available in small research pack sizes (25 mg, 50 mg, 100 mg).
  • Chemical formula C16H12N2O5; Mw 312.28 g·mol⁻¹.

Catalog No. 50-002-81095


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.