Learn More
Apexbio Technology LLC Thalidomide 50-35-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000094020
Encompass_Preferred
Thalidomide (CAS 50-35-1) is a small-molecule inhibitor targeting the CRBN-DDB1-Cul4A E3 ubiquitin ligase complex It is designed to modulate ubiquitination pathways thereby regulating protein degradation cellular proliferation and immune signaling Thalidomide exerts its biological activity primarily through inhibition of the CRBN-DDB1-Cul4A complex In cell-based studies thalidomide demonstrates inhibitory activity with reported IC50 values ranging from approximately 1 M to 100 M depending on the assay and cell line Based on these pharmacological properties thalidomide holds research potential in the fields of immune regulation angiogenesis inhibition and cancer biology particularly in studies of hematological malignancies such as multiple myeloma
Catalog No. 50-000-94020 Supplier Apexbio Technology LLC Supplier No. A42165.1
Thalidomide (CAS 50-35-1) is a small-molecule inhibitor targeting the CRBN-DDB1-Cul4A E3 ubiquitin ligase complex It is designed to modulate ubiquitination pathways thereby regulating protein degradation cellular proliferation and immune signaling Thalidomide exerts its biological activity primarily through inhibition of the CRBN-DDB1-Cul4A complex In cell-based studies thalidomide demonstrates inhibitory activity with reported IC50 values ranging from approximately 1 M to 100 M depending on the assay and cell line Based on these pharmacological properties thalidomide holds research potential in the fields of immune regulation angiogenesis inhibition and cancer biology particularly in studies of hematological malignancies such as multiple myeloma