Learn More
Cayman Chemical ThalIdomIde 5mg
SDP
Catalog No. 502970177
Encompass_Preferred
An active enantiomer of (±)-thalidomide; enhances TPA-induced TNF-α production in HL-60 cells from 0.1-100 µM; preincubation of isolated RIR chicken donor blood reduces recipient chicken embryo splenomegaly in a model of graft versus host disease at 10 mg/ml; induces fetal malformations when administered to pregnant female rabbits at 150 mg/kg
Catalog No. 50-297-0177 Supplier Cayman Chemical Supplier No. 149675 MG
An active enantiomer of (±)-thalidomide; enhances TPA-induced TNF-α production in HL-60 cells from 0.1-100 µM; preincubation of isolated RIR chicken donor blood reduces recipient chicken embryo splenomegaly in a model of graft versus host disease at 10 mg/ml; induces fetal malformations when administered to pregnant female rabbits at 150 mg/kg